missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS, 95.0-98.0%

Product Code. 11327588
Change view
Click to view available options
Quantity:
1 kg
4 x 1 kg
Dimensione della confezione:
1kg
4kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11327588 1 kg 1kg
11337588 4 x 1 kg 4kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativa: Questo prodotto contiene un precursore di esplosivi ai sensi del regolamento (CE) n. 1148/2019. Ogni cliente deve compilare una dichiarazione prima della consegna.
Codice prodotto. 11327588 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 033273.A1
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativa: Questo prodotto contiene un precursore di esplosivi ai sensi del regolamento (CE) n. 1148/2019. Ogni cliente deve compilare una dichiarazione prima della consegna.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid is used as an electrolyte in lead-acid batteries, and a cleaning agent. It is used in mineral processing, fertilizer manufacturing, oil refining, wastewater processing, and chemical synthesis. It is also used in the production of detergents, synthetic resins, dyestuffs, pharmaceuticals, petroleum catalysts, antifreeze, processes such as oil well acidizing, aluminum reduction, paper sizing, pigments, enamels, printing inks, coated fabrics and paper. It is also used in the production of explosives, cellophane, acetate and viscose textiles, lubricants, non-ferrous metals and batteries. Sulfuric acid immobilized on silica gel is used as an efficient and benign catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifica

Density 1.84
Assay Percent Range 95.0 to 98.0%
Quantity 1 kg
Physical Form Liquid
Solubility Information Sulfuric acid is miscible with water at all concentrations.
Formula Weight 98.08
Grade ACS
Chemical Name or Material Sulfuric Acid
compliance-icons
Identificativo prodotto
  • Sulfuric acid, ACS, 95.0-98.0%
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consigli di prudenza
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contain: sulphuric acid

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.