missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS reagent, 95% solution in water

Product Code. 10351741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass Bottle
HDPE Bottles
Plastic bottle
Plastic drum
Unit Size:
10 L
2.5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351741 500 mL Glass Bottle 500mL
10391313 500 mL Plastic bottle 500mL
10015910 2.5 L HDPE Bottles 2.5 L
10536933 2.5 L Plastic bottle 2.5 L
11914091 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10351741 Supplier Thermo Scientific Acros Supplier No. 424525000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point 10.0°C
Boiling Point 290.0°C
Density 1.8300g/mL
Heavy Metals (as Pb) 1ppm max.
Packaging Glass Bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.83
Assay Percent Range 95%
Quantity 500 mL
Viscosity 21 mPa.s (25°C)
Linear Formula H2SO4
Physical Form Liquid Free From Suspended or Insoluble Matter
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Percent Purity 95.0 to 98.0%
Grade ACS Reagent
Chemical Name or Material Sulfuric acid
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.