missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, for analysis, ca. 96% solution in water

Product Code. 10765663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10765663 1 L Plastic bottle 1 L
10624792 500 mL Glass Bottle 500mL
10699824 500 mL Plastic bottle 500mL
10244790 1 L Glass Bottle 1 L
10011240 2.5 L Glass Bottle 2.5 L
10322963 2.5 L Plastic bottle 2.5 L
11904091 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10765663 Supplier Thermo Scientific Acros Supplier No. 124640011
  • ONLINE EXCLUSIVE PRICE
    51.40 €
    47.50 € / 1 L
    Save 3.90 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point 10.0°C
Color Colorless
Density 1.8300g/mL
Boiling Point 290.0°C
Ignition Residue 0.0005% max.
Packaging Plastic bottle
Linear Formula H2SO4
Quantity 1 L
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Specific Gravity 1.83
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Percent Purity 95 to 97%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Sulfuric acid
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.