missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, Environmental Grade, 93-98%

Product Code. 11339027
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339027 2.5 L 2.5 L
11440191 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11339027 Supplier Thermo Scientific Alfa Aesar Supplier No. 038751.K4
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid has a strong dehydrating agent and is applied in many applications like the domestic acidic drain cleaner, electrolyte in lead-acid batteries and in various other cleansing agents. It is also useful for mineral processing, fertilizer manufacturing, petroleum purification, wastewater processing, and chemical synthesis. The catalytic decomposition of sulfuric acid when mixed with Cr/Fe oxides, shows a useful application in the sulfur iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifikationer

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.184 g/cm3 at 20°C
Odor Acrid
Assay Percent Range 93 - 98%
Quantity 2.5 L
Physical Form Liquid,single sub-boiling quartz distillation
UN Number 1830
Solubility Information Miscible in water.
Formula Weight 98.08
Grade Environmental
Chemical Name or Material Sulfuric acid, Environmental Grade
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Sulfuric acid, Environmental Grade, 93-98%
Signalord
  • Danger
Riskkategorier
  • Skin corrosion/irritation Category 1 A
Faroangivelser
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.