missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Bag in box |
| Vapor Pressure | 1mmHg at 20°C |
| Quantity | 10 L |
| Physical Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?