missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution, Ready to Use, for Volumetric Analysis, meets analytical spec of Ph.Eur., BP, Fisher Chemical™

Product Code. 10418030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418030 2.5 L HDPE plastic bottle 2.5 L
12688635 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10418030 Supplier Fisher Chemical Supplier No. J/8430/17
  • ONLINE EXCLUSIVE PRICE
    58.45 €
    56.90 € / 2.5 L
    Save 1.55 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

Sulfuric Acid Solution 0.5M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 1.5
Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Vapor Pressure 1mmHg at 20°C
Quantity 2.5 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Sulfuric acid, solution 0.5M, (1N)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Sulfuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.