missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N), Honeywell Fluka™

Product Code. 15694980 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC Container
Unit Size:
1 L
10 L
2.5 L
5 L
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15694980 6 x 1 L Plastic Bottle Pack of 6
15634980 1 L Plastic Bottle 1 L
15644980 2.5 L Plastic Bottle 2.5 L
15674980 5 L Plastic Bottle 5 L
15624980 10 L VOLPAC Container 10 L
15654980 4 x 2.5 L Plastic Bottle Pack of 4
15614980 10 L Plastic Bottle 10 L
15664980 4 x 5 L Plastic Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15694980 Supplier Honeywell-Fluka Supplier No. 353546X1L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 100°C
CAS Min % ≥90.0000%, ≥1.0000%
Concentration 0.5 M H2SO4 (1.0N)
CAS Max % ≤100.0000%, <5.0000%
Density 1.030 g/cm3
Packaging Plastic Bottle
Quantity 6 x 1 L
Linear Formula H2SO4
UN Number UN3264
Beilstein 2037554
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Formula Weight 98.08
Grade Reagent Ph. Eur.
Chemical Name or Material Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N)
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric acid solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.