missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution, volumetric, 0.25 M H2SO4 (0.5N), Honeywell™ Fluka™

Product Code. 15613590 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
Unit Size:
2 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15613590 500 mL Poly Bottle 500mL
15603590 2 L Poly Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15613590 Supplier Honeywell-Fluka Supplier No. 319570500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point >100°C
Density 1.01g/mL at 25°C
Packaging Poly Bottle
Quantity 500 mL
Linear Formula H2SO4
UN Number UN3264
Beilstein 2037554
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric Acid Solution, Volumetric, 0.25M H2SO4 (0.5N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.