missing translation for 'onlineSavingsMsg'
Learn More

Sulfurous acid, ACS, 6.0% SO{2} min

Product Code. 11444990
Change view
Click to view available options
Quantity:
500 mL
4 x 500 mL
Unit Size:
2000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444990 4 x 500 mL 2000mL
11434990 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444990 Supplier Thermo Scientific Alfa Aesar Supplier No. 033275.AY
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7782-99-2 | H2O3S | 82.073 g/mol

Sulfurous acid is used as a reducing agent, bleaching agent and disinfectant. It is utilized for bleaching of moistened wool, straw for hats, and other materials injured by chlorine. It finds application as a disinfectant as well as used externally for its parasiticidal effect in various skin diseases. Its derivative viz. sulfites are included in antiseptics, antifermentatives, and antizymotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-99-2
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.073
MDL Number MFCD00011335
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid
PubChem CID 1100
ChEBI CHEBI:48854
IUPAC Name sulfurous acid
SMILES OS(=O)O

Specifications

Appearance Colourless liquid
Color Colorless
Density 1.03 g/cm3 at 20°C
Odor Pungent
Sensitivity Air Sensitive
Quantity 4 x 500 mL
Physical Form Liquid
UN Number 1833
Solubility Information Miscible with water.
Formula Weight 82.08
Grade ACS
Chemical Name or Material Sulfurous Acid, 6.0% SO2 min.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulphurous acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sulfurous acid

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.