missing translation for 'onlineSavingsMsg'
Learn More

Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™

Product Code. 10433672 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10433672

Brand: Thermo Scientific Acros 378151000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfuryl chloride
Name Note 1.0M Solution in Dichloromethane
CAS 7791-25-5, 75-09-2
Quantity 100 mL
Molecular Formula Cl2O2S
Linear Formula SO2Cl2
MDL Number MFCD00011451
Merck Index 15, 9110
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
SMILES ClS(Cl)(=O)=O
IUPAC Name sulfuryl dichloride
Molecular Weight (g/mol) 134.96
PubChem CID 24648
ChEBI CHEBI:29291
Formula Weight 134.97
Concentration or Composition (by Analyte or Components) 1M, exact strength on the certificate of analysis
Packaging AcroSeal™ Glass bottle
Appearance Clear pale yellow solution
Specific Gravity 1.352
Density 1.3520g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sulfuryl chloride, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H331-Toxic if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.