missing translation for 'onlineSavingsMsg'
Learn More

Sulisobenzone, TRC

Product Code. 30138420
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138420 10 g 10g
30137352 5 g 5g
30007983 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138420 Supplier Toronto Research Chemicals Supplier No. TRCS78947010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4065-45-6
Molecular Formula C14 H12 O6 S
Molecular Weight (g/mol) 308.31
Synonym 1-Phenol-4-sulfonic acid, 2-benzoyl-5-methoxy- (6CI), 5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, 2-Hydroxy-4-methoxy-5-sulfobenzophenone, 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, 2-Methoxy-4-hydroxy-5-benzoylbenzenesulfonic acid, 3-Benzoyl-4-hydroxy-6-methoxybenzenesulfonic acid, Benzophenone 4, Escalol 577, MS 40, NSC 60584, Seesorb 101S, Spectra-Sorb UV 284, Sulisobenzone, Sungard, Syntase 230, UV 284, Uval, Uvasorb S 5, Uvinul D 5030, Uvinul MS 40, Viosorb 111
IUPAC Name 5-benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid
SMILES COc1cc(O)c(cc1S(=O)(=O)O)C(=O)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H12O6S/c1-20-12-8-11(15)10(7-13(12)21(17,18)19)14(16)9-5-3-2-4-6-9/h2-8,15H,1H3,(H,17,18,19)
Formula Weight 308.0355
Percent Purity >95%
Chemical Name or Material 5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.