missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid Concentrated Volumetric Solution Ampule, Reagecon
BIOPHARMA

Product Code. 11576713 Shop All Reagecon Products
Click to view available options
:
1 Ampule
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11576713

Brand: Reagecon SUC051L

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Fully factorized, high purity, stable product, developed and tested for titrations.

  • Use to make 1 L of 0.5 M (1.0 N) Sulphuric Acid
  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • When the product is accurately diluted to one liter in a volumetric flask, it can then be used as an analytical volumetric solution, to a specification of ± 0.2%.
  • The product must be diluted accurately to one liter, in a volumetric flask under controlled temperature conditions, for optimal performance.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.

Specifications

Concentration 0.5 M (1.0 N)
Packaging Ampule
Quantity 1 Ampule
Physical Form Liquid
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.