missing translation for 'onlineSavingsMsg'
Learn More

Sulphuric Acid Solution, 0.5M, Fisher Chemical™
BIOPHARMA

Product Code. 12943634 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12943634 1000 mL 1000mL
12920774 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12943634 Supplier Fisher Chemical Supplier No. VL 317 1000
  • ONLINE EXCLUSIVE PRICE
    42.60 €
    38.98 € / 1000mL
    Save 3.62 €
    8% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 0.5, 1mol/L, eq/L
Quantity 1000 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.