missing translation for 'onlineSavingsMsg'
Learn More

Sumatriptan, TRC

Product Code. 30132766
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132766 1 g 1g
30135134 250 mg 250mg
17971545 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132766 Supplier Toronto Research Chemicals Supplier No. TRCS8000951G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 103628-46-2
Molecular Formula C14 H21 N3 O2 S
Molecular Weight (g/mol) 295.4
Synonym 3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide, 1-[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide, 3-[2-(Dimethylamino)ethyl]-N-methylindolyl-5-methanesulfonamide, GR 43175, GR 43175X, Onzetra Xsail, Sumatriptan, Zecuity, Zelrix
IUPAC Name 1-[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]-N-methylmethanesulfonamide
SMILES CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)C)c2c1

Specifica

Quantity 1 g
InChI Formula InChI=1S/C14H21N3O2S/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3/h4-5,8-9,15-16H,6-7,10H2,1-3H3
Formula Weight 295.135
Percent Purity >95%
Chemical Name or Material Sumatriptan
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.