missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Super Bright Complete Staining Buffer, eBioscience

Product Code. 16349553
Change view
Click to view available options
Quantity:
100 tests
1000 tests
Unit Size:
Pack of 100
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16349553 100 tests Pack of 100
16359553 1000 tests Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16349553 Supplier Invitrogen™ Supplier No. SB440142
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Super Bright Complete Staining Buffer, eBioscience, is designed for use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension.

Super Bright Complete Staining Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample to prevent non-specific polymer interactions, which can result in data appearing under-compensated. Super Bright Complete Staining Buffer is provided in a convenient 5 μL/test format and is compatible with traditional fluorochromes, Brilliant Violet dyes, and standard flow cytometry protocols.

Super Bright Complete Staining Buffer is not compatible with UltraComp eBeads microspheres. If using UltraComp eBeads microspheres as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres.

Super Bright Complete Staining Buffer is a fully compatible replacement for Super Bright Staining Buffer (Cat. Nos. SB-4400-42, SB-4400-75), and it shows decreased background in the 450/40 channel on the violet laser.

Super Bright polymer dyes are sold under license from Becton, Dickinson and Company.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 0.5 mL, store at 2°C to 8°C
Quantity 100 tests
Volume (Metric) 0.5 mL
Color Black
Shipping Condition Wet Ice
For Use With (Application) Flow Cytometry
Product Line eBioscience
Product Type Staining Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.