Learn More
uvex Super F OTG Clear SV EXC Overspecs, Black
Product Code. 31751180 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751180 Polycarbonate Each
1 options
Product Code. 31751180 Supplier uvex Supplier No. 9139265
  • ONLINE EXCLUSIVE PRICE
    20.19 €
    19.35 € / Pack of 1
    Save 0.84 €
    4% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provide optimum protection and high compatibility with prescription spectacles, featuring a large lens for excellent visibility and comfort.
  • Modern uvex OTG Plus overspecs designed for prescription spectacle wearers, featuring an extremely large, flat clear SV EXC lens that offers excellent vision and sufficient space to comfortably wear even large prescription glasses underneath.
  • Metal-free black frame construction with flat, flexible side arms and a tilt-adjustable lens to ensure maximum comfort and individual adaptability.
  • Innovative frame with multi-incline adjustment points allows a high level of individualisation for a secure fit across different facial shapes and head widths.
  • Ergonomically shaped, flat side arms with soft-grip ear-pieces provide a comfortable, secure fit without pressure points, complemented by an anatomically designed nose piece for all-day comfort and optimal compatibility with respirators.
  • Certified according to EN ISO 16321-1 for personal eye and face protection in industrial use, featuring a robust polycarbonate lens with proven uvex supravision coating technology that is extremely scratch-resistant on both sides.
  • UV400 protection delivering 100% protection against dangerous ultraviolet radiation up to 400 nm.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Overglasses
Frame Color Black
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Adjustable Temples
Autoclavable Not Autoclavable