missing translation for 'onlineSavingsMsg'
Learn More

SICCO Super-Maxi 1-Desiccator

Product Code. 10679464
Change view
Click to view available options
Volume (Metric):
311 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10679464 311 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679464 Supplier SICCO Supplier No. V193201
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Super-Maxi 1-Desiccator ACRYL/ALUMINIUM, with elastic gasket-lined door, 1 desiccant tray, 1 mounted hygrometer, 1 magnetic lock, 4 removable shelves made of stainless steel, 2 quick couplings for gas-filling and connecting tube (length 5 m), 4 casters (2 of them with brakes). Usable interior space: W 495 x H 1030 x D 540 mm

  • Aluminium frame with panels made of acrylic glass, including four shelves made of stainless steel, tray, hygrometer and hose (5 m) with two quick couplings for gas filling, four casters, usable with up to 17 shelves.
  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch, circular rubber seal
  • Extra big electronic hygrometer
  • Variable height shelves
  • Two connections for gas filling including quick couplings with self-sealing valves and hose
  • Telescopic shelves

Specifications

Cabinet Style Front-loading
Product Type Nonvacuum Desiccator
Volume (Metric) 311 L
Dimensions (D x W x H) Interior 495 x 1030 x 540 mm
Weight (Metric) 30 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.