Learn More
Hamilton™ Super Syringe Gastight™ Syringes
Product Code. 10379837
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
1.5 L
2000 mL
Includes:
No needle available
No needle included
Model:
S0500
S1000
S1500
S2000
Connection Type:
Luer Lock
Tracheal Adapter
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes Model Connection Type Unit Size
10379837 1000 mL No needle included S1000 Luer Lock Each
11587884 500 mL No needle available S0500 Tracheal Adapter Each
10885780 2000 mL No needle included S2000 Luer Lock Each
11577884 500 mL No needle included S0500 Luer Lock Each
11547984 1.5 L No needle included S1500 Luer Lock Each
11523524 1000 mL No needle available S1000 Tracheal Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10379837 Supplier Hamilton™ Supplier No. 86312
  • ONLINE EXCLUSIVE PRICE
    1350.00 €
    1244.72 € / Pack of 1
    Save 105.28 €
    7% Off
View all promotions
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Large volume GASTIGHT syringe ideal for dispensing volumes from 50mL up to 2L. The Super Syringe is the only Hamilton syringe that is made with an acrylic barrel instead of a glass barrel.
Super Syringe Terminations
  • Trachael- This termination accepts 5/8 inch inner diameter flexible tubing
  • PTFE Luer Lock (TLL) - This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25mL and greater syringes. Repeated autoclaving will shorten syringe life.
Needle Point Styles (Point styles are available on Kel-f Hub and Metal Hub Needles)
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 33-10
  • Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 33-10. Special point styles such as 30°, 45°, or any other angle are available upon request
  • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
  • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 26 & 22
† No Needle Available
Specifications
Material Acrylic (Barrel)
For Use With (Application) For air sampling, preparing gas standards, calibrating reservoirs, pneumographs and spirometers
Includes No needle included
Product Type General Purpose Syringe
Model S1000
Volume (Metric) 1000 mL
Connection Type Luer Lock