missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SuperFastPrep-2™ Portable Bead Beating Grinder and Lysis System, 2 x 2 mL, Battery Operated

Product Code. 15413654 Shop All MP Biomedicals Products
Change view
Click to view available options
Sample Volume:
Up to 200 mg in 1 mL buffer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Volume unitSize
15413654 Up to 200 mg in 1 mL buffer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413654 Supplier MP Biomedicals™ Supplier No. 116012500
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

SuperFastPrep-2™ Portable Bead Beating Grinder and Lysis System is used for Sample Preparation, lysis, homogenization and grinding of virtually any type of biological sample. It is handheld and compatible with most 2 mL tubes containing lysis matrix.

  • Product Overview: The SuperFastPrep2 is a compact, cordless, hand-held sample disruption instrument designed to quickly and efficiently homogenize, grind, or lyse biological and environmental samples. It can be used at remote locations and in most weather conditions for the immediate collection, processing, and preservation of samples.
  • SuperFastPrep-2 uses a unique mechanism mounted on a rotary tool designed to operate at 600 to 4,800 cycles per minute with a fully charged battery. The self-contained device holds two standard 2 mL screw cap Lysing Matrix tubes optimized for a wide selection of sample types. Included with the unit is a rotary handle, two lithium ion rechargeable batteries, charging station, voltage converter, and protective carrying case.
  • Applications: Used to homogenize, grind or lyse any samples, the SuperFastPrep-2 portable homogenizer thoroughly and quickly lyses any tissue and cell type, allowing easy and reproducible isolation of stable RNA, active proteins and full-length genomic DNA.
  • Sample Volume: Up to 200 mg in 1 mL buffer
  • Capacity: 2 x 2 mL Tube
  • 90/240 V for lab version, cordless operation for field version.

Recommended Storage

Store at Room Temperature (15–30°C).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) It can be used to lyse microbes in soil, sediment, sludge, and fecal samples and can effectively process tough-to-lyse fungal, algal, plant, and animal tissues. It can be used at any remote location and in most weather conditions when immediate sample collection, processing, and preservation are required by the researcher.
Noise Level 80 to 100 dB
Sample Volume Up to 200 mg in 1 mL buffer

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.