missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ SuperFrost™ Microscope Slides

Product Code. 17204894
Change view
Click to view available options
Color:
Blue
Green
Pink
Yellow
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17204894 Pink Pack of 50
17294884 Blue Pack of 50
17214894 Yellow Pack of 50
17224894 Green Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17204894 Supplier Epredia™ Supplier No. AG00008232E01MNZ20
  • ONLINE EXCLUSIVE PRICE
    8.63 €
    4.66 € / Pack of 50
    Save 3.97 €
    46% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors allows several examination specimens to be labeled and archived simply and clearly.

  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
TRUSTED_SUSTAINABILITY

Specifications

Edges 90°, Ground
Thickness (Metric) 1 mm
Color Pink
Certifications/Compliance ISO8037/I
Coating Uncoated
Writing Surface Yes
Dimensions (L x W) 76 x 26 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.