Learn More
Heinz Herenz Superfrost Slides Ground Edges
Click to view available options
:
With Plain Corner Style
Description
We offer a very wide range of slides, so you are sure to find what you are looking for.
- Within the product lines you will find cut and ground slides.
- Cut slides are preferred for routine microscopy and offer great flexibility.
- Ground slides prevent cuts due to the ground edges.
- If you want to label and categorize your slides, it is best to choose slides with matted edges or a coloured field.
- You also have the choice between slides with and without cellophane.
Specifications
Specifications
| Sterility | Non-sterile |
| Edges | 90°, Ground |
| Frosted | SuperFrost |
| Material | Glass |
| Precleaned | No |
| Type | Plain Microscope Slide |
| Well Count | 0 |
| Color | Transparent |
| Corner Style | Plain |