missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Superose™ 6 Increase Columns

Product Code. 15377653 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.2 mm
5 mm
10 mm
Height (Metric):
150 mm
300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Height (Metric) Unit Size
15377653 10 mm 300 mm Each
15303234 3.2 mm 300 mm Each
15383224 5 mm 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15377653 Supplier Cytiva™ Supplier No. 29091596
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-resolution size-exclusion chromatography for studying large proteins and complexes. Cytiva Lifescience™ Superose™ 6 Increase Columns delivers high quality in size exclusion chromatography (SEC).

Superose™ 6 Increase pre-packed columns deliver the quality and high resolution in size exclusion chromatography (SEC) — even for large proteins and aggregates.

Increased performance

  • Higher resolution than traditional Superose 6 columns
  • Wide fractionation range (Mr from 5,000 to 5,000,000)
  • Withstands high-pH cleaning
TRUSTED_SUSTAINABILITY

Specifications

Product Type Superose 6 Increase 10/300 GL
Diameter (Metric) Inner 10 mm
Height (Metric) 300 mm
Flow Rate 0.5mL/min.
Material Borosilicate glass
Ligand Type Cross-linked agarose
Max. Pressure 435 psi
pH Range 3 to 12
Particle Size 8.6μm
Support Type Polyethylene (PE) filter
Content And Storage 4° to 30° in 20% ethanol
Column Type Gel Filtration Column
Show More Show Less
compliance-icons
Product Identifier
  • Superose 6 Increase 10/300 GL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.