missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV CellsDirect™ cDNA Synthesis Kit
GREENER_CHOICE

Product Code. 16653873
Change view
Click to view available options
No. of Reactions:
50 Reactions
500 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
16653873 500 Reactions Each
16643873 50 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16653873 Supplier Invitrogen™ Supplier No. 11750350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The SuperScript IV CellsDirect cDNA Synthesis Kit is optimized for the synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating RNA. Cell lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in PCR and qPCR.

The SuperScript IV CellsDirect cDNA Synthesis Kit is optimized for the synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating RNA. Cell lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in PCR and qPCR.

Features of the SuperScript IV CellsDirect Synthesis Kit
• Compatible with a wide range of mammalian cell types grown under different treatment conditions
• Single-tube format minimizes reagent loss, sample loss, and handling time
• Total lysate volume can be used in first-strand cDNA synthesis reaction, providing greater yields with a limited number of cells and allowing for detection of rare transcripts
• High-quality cDNA generated for use in a variety of applications, including PCR and qPCR
• Simple protocol takes approximately 37 minutes
• Compatible with TaqMan and SYBR Green qPCR master mixes from various manufacturers
• Optimized performance for 1–10,000 cells per sample, results equivalent to those from purified RNA

Features of the SuperScript IV Reverse Transcriptase included in this kit
• Reduced RNase H activity
• High thermal stability
• High yields of cDNA in the first-strand synthesis reaction for greater sensitivity and enhanced detection of rare transcripts
• Superior tolerance to various RT inhibitors

SuperScript IV reverse transcriptase is a top choice for all RT-PCR and qRT-PCR applications. It is a proprietary MMLV mutant with superior robustness and reliability in RT reactions. It is significantly improved over SuperScript III reverse transcriptase in inhibitor resistance, processivity, and reaction speed, while retaining all the benefits of the previous enzyme, including increased thermostability, highly efficient full-length cDNA synthesis, and reduced RNase activity.

How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript IV CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I treatment is an optional step and can be used to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• SuperScript IV CellsDirect Lysis Solution (3 x 9 mL)
• SuperScript IV CellsDirect Stop Solution (2 x 1.25 mL)
• Lysis Enhancer (1 x 250 μL)
• DNase I (1 x 500 μL)
• SuperScript IV RT Master Mix (4 x 1 mL)
• SuperScript IV No RT Control (4 x 1 mL)

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 10 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity 500 reactions
Reverse Transcriptase SuperScript IV
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) RT-PCR, Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 500 Reactions
Reaction Format Premixed Components
Reagent Type Reverse Transcription
Starting Material Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.