missing translation for 'onlineSavingsMsg'
Learn More

Captopril Disulfide, TRC

Product Code. 18090010
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18090010 10 mg 10mg
18001120 5 mg 5mg
18021920 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18090010 Supplier Toronto Research Chemicals Supplier No. TRCC17576010MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64806-05-9
Molecular Formula C18 H28 N2 O6 S2
Molecular Weight (g/mol) 432.55
Synonym 1,1'-[Disulfanediylbis-[(2S)-2-methyl-1-oxopropane-3,1-diyl]]-bis[(2S)-pyrrolidine-2-carboxylic] Acid, Captopril Imp. A (EP), Captopril Disulfide
IUPAC Name (2S)-1-[(2S)-3-[[(2S)-3-[(2S)-2-carboxypyrrolidin-1-yl]-2-methyl-3-oxopropyl]disulfanyl]-2-methylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES C[C@H](CSSC[C@@H](C)C(=O)N1CCC[C@H]1C(=O)O)C(=O)N2CCC[C@H]2C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H28N2O6S2/c1-11(15(21)19-7-3-5-13(19)17(23)24)9-27-28-10-12(2)16(22)20-8-4-6-14(20)18(25)26/h11-14H,3-10H2,1-2H3,(H,23,24)(H,25,26)/t11-,12-,13+,14+/m1/s1
Formula Weight 432.1389
Percent Purity >95%
Chemical Name or Material Captopril Disulfide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.