missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ Western Blot Enhancer

Product Code. 10157958 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10157958 500 mL Each
10801550 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157958 Supplier Thermo Scientific™ Supplier No. 46640
Protein Biology Essentials
Use promo code 27946
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Increase both signal intensity and sensitivity 3- to 10-fold with this membrane treatment reagent and primary antibody diluent.

Thermo Scientific™ SuperSignal Western Blot Enhancer contains a membrane treatment reagent and a primary antibody diluent that increase both signal intensity and sensitivity 3- to 10-fold compared to detection performed without it.

When a protein or antigen is difficult to detect because of low abundance or poor immunoreactivity, use of SuperSignal Western Blot Enhancer can significantly reduce background and enhance detection of low-abundance and weakly immunoreactive antigens. This versatile kit works with PVDF and nitrocellulose membranes and is compatible with fluorescent, chromogenic and chemiluminescent detection.

Highlights:

  • Increase sensitivity – achieve 3- to 10-fold increase in signal intensity and sensitivity

  • Improve specificity – improves signal-to-noise ratio for poor quality and low affinity antibodies

  • Better clarity – reduces background for cleaner Western blots

  • Membrane compatibility – provides effective signal enhancement with PVDF and nitrocellulose membranes

  • Substrate compatibility – validated for use with chromogenic, chemiluminescent and fluorescent detection methods

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Enhancer
Detection Method Chemiluminescence, Fluorescence, Colorimetric
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 500 mL
Content And Storage Upon receipt store solutions at 4°C Products are shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.