missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ Western Blot Substrate Bundle, Atto + trial-size Pico PLUS

Product Code. 17156378 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL + 20 mL bundle
Unit Size:
120mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17156378 100 mL + 20 mL bundle 120mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17156378 Supplier Thermo Scientific™ Supplier No. A45917
  • ONLINE EXCLUSIVE PRICE
    526.00 €
    426.06 € / 120mL
    Save 99.94 €
    19% Off
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains SuperSignal™ West Atto Ultimate Sensitivity Chemiluminescent Substrate plus a sample size of SuperSignal™ West Pico PLUS Chemiluminescent Substrate to provide chemiluminescent detection substrates for both routine and high sensitivity western blotting.

  • SuperSignal West Atto
    • Sensitivity: low-femtogram to high-attogram
    • Best for low abundant targets or precious samples
  • SuperSignal West Pico PLUS
    • Picogram to femtogram sensitivity: detect low-picogram to high-femtogram amounts of target protein on nitrocellulose or PVDF membrane
    • Best for routine western blots
TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 100 mL + 20 mL bundle
Substrate Horseradish Peroxidase
Sensitivity Picogram to femtogram, low-femtogram to high-attogram
Sufficient For 12 Mini-Gel Blots, 1000 sq. cm of Membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane
Content And Storage • 100 mL SuperSignal West Atto reagent, store at 2–8°C
• 20 mL SuperSignal West Pico PLUS reagent, store at room temperature

Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.