missing translation for 'onlineSavingsMsg'
Learn More

SuperSignal™ Western Blot Substrate Bundle, Femto + trial-size Pico PLUS

Product Code. 17153209 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 mL bundle
Unit Size:
200mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17153209 200 mL bundle 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17153209 Supplier Thermo Scientific Supplier No. A43841
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

This substrate kit is comprised of SuperSignal West Femto Maximum Sensitivity Substrate plus a sample size of SuperSignal West Pico PLUS Chemiluminescent Substrate to provide chemiluminescent detection substrates for both high sensitivity and routine western blotting.

SuperSignal West Femto Maximum Sensitivity Substrate

Thermo Scientific SuperSignal West Femto Maximum Sensitivity Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for low femtogram protein level detection by western blot analysis with horseradish peroxidase (HRP) enzyme. It is optimized to detect low abundant and/or difficult to detect targets.

  • Sensitivity: low-femtogram (mid-zeptomole)
  • Stability: 8-hr working solution stability, 1-year kit stability at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to 8 hours
  • Recommended primary antibody concentration: 1:5,000–1:100,000 dilution (10–200 ng/mL)
  • Recommended secondary antibody concentration: 1:100,000–1:500,000 dilution (2–10 ng/mL)

SuperSignal West Pico PLUS Chemiluminescent Substrate

Thermo Scientific SuperSignal West Pico PLUS Chemiluminescent Substrate enables picogram to high femtogram-level protein detection by western blot analysis. It is best for routine, everyday western blotting applications, providing excellent signal intensity, sensitivity and duration across a wide range of targets.

  • Sensitivity: pictogram to femtogram sensitivity
  • Stability: 8-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 6 to 24 hours
  • Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:20,000–100,000 dilution (10–50 ng/mL)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 200 mL bundle
Substrate Horseradish Peroxidase
Sensitivity Picogram to femtogram, low femtogram (mid-zeptomole)
Sufficient For 60 Mini-Gel Blots, 5000 sq. cm of membrane; 2 Mini-Gel Blots, 200 sq. cm of Membrane
Content And Storage • 200 mL SuperSgnal West Femto reagent, store at 2–8°C
• 20 mL SuperSignal West Pico PLUS reagent, store at room temperature

Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.