missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ Heavy-duty Disposable Nitrile Gloves

Product Code. 15269363
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15269363 Medium Pack of 100
15259363 Large Pack of 100
15279363 Small Pack of 100
15289363 X-Large Pack of 100
15299363 X-Small Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15269363 Supplier Ansell Supplier No. 93843080
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides protection while assisting workers with improved efficiency and speed. Ansell™ MICROFLEX™ Heavy-duty Disposable Nitrile Gloves resist tape and adhesives with a low pinhole acceptance rate of 0.65 AQL.

  • Offers an advanced barrier protection against contaminants
  • Non-stick properties are built into the glove formulation to create a resistance to tape or adhesives
  • Textured fingers ensure strong, efficient grip and maximum worker protection
  • Polymer coating assures easy donning to help protect workers in fast-paced environments
  • Constructed without natural rubber latex materials or powder, to help protect from allergies, skin irritation and dryness
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN-374-2 Level 3, EN-374, 93/42/EEC, EN-455 Parts 1, 2 and 3, ASTM F1671
Accelerator Free No
Applications Industrial
AQL 0.65
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.11 mm
Finger Style Textured
For Use With (Application) Healthcare, dental, industrial safety, laboratory, EMS
Glove Cuff Style Beaded/Rolled
Model 93-843
Packaging Type Dispenser Box
Quantity 100/Pk.
Tensile Strength 14 to 30 MPa
Length (Metric) 24.5 cm
Width (Metric) 98 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.