missing translation for 'onlineSavingsMsg'
Learn More

Microflex™ High Risk Disposable Nitrile Gloves

Product Code. 15267049
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15267049 X-Large Pack of 50
15247049 Medium Pack of 50
15257049 Large Pack of 50
15237049 Small Pack of 50
15503705 2X-Large Pack of 50
15523705 X-Small Pack of 50
15656054 3X-Large Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15267049 Supplier Microflex Supplier No. 93853100
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offer up to 3 times the puncture resistance of latex or vinyl disposable gloves. Microflex™ High Risk Disposable Nitrile Gloves also provide chemical and liquid protection.

  • Extended cuff provides additional protection for the wrist and arm for hazardous environments.
  • Advanced nitrile formulation provides the lowest acceptable pinhole rates (AQL 0.65)
  • Non-stick properties are built into the glove formulation to create a resistance to tape or adhesives, and to assist workers with improved efficiency and speed.
  • Textured fingers ensure strong, efficient grip and excellent worker protection.
  • Constructed without natural rubber latex materials or powder to help protect from allergies, skin irritation and dryness.
  • Approved for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, ISO 11193-1, EN420, 89/686/EEC, EN374-2: 2003, EN374-3: 2003, MDD 93/42/EEC, Class 1, EN455-1, EN455-2 and EN455-3, ASTM F1671
Applications Industrial
Color Blue
Disposable Single-use
Finger Thickness (English) 0.0087 in.
Finger Thickness (Metric) 0.22 mm
Grade Medical
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0055 in.
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Product Type Chemical-resistant Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 0.22 mm
Thickness (Metric) 0.14 mm
Finger Style Textured
Glove Cuff Style Extended, Beaded
Model 93-853
Packaging Type Dispenser Box
Quantity 50Pack
Tensile Strength 14 to 30 MPa
Length (Metric) 295 mm
Width (Metric) 115 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.