missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Suramin hexasodium salt

Product Code. 30040211
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040211 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040211 Supplier R&D Systems™ Supplier No. 1472/50
  • ONLINE EXCLUSIVE PRICE
    136.08 €
    120.75 € / 50mg
    Save 15.33 €
    11% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective P2 antagonist

Suramin hexasodium salt is a non-selective P2 purinergic antagonist. Also blocks calmodulin binding to recognition sites and G protein coupling to G protein-coupled receptors. Increases open probability of ryanodine receptor (RyR) channels and acts as a competitive α1β2γ2 GABAA receptor antagonist. Anticancer, antiviral and antiparasitic agent.

This product may be supplied with a high degree of hydration and some residual NaCl, the amounts of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8,8'-[Carbonylbis[imino-3,1-phenylenecarbonylimino(4-methyl-3,1-phenylene)carbonylimino]]bis-1,3,5-naphthalenetrisulfonic acid hexasodium salt
CAS 129-46-4
Quantity 50 mg
Product Line Tocris
Target P2X Receptor Antagonists
Molecular Formula C51H34N6Na6O23S6
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.