missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Glass Plates

Product Code. 15593371
Change view
Click to view available options
Quantity:
2 sets
Unit Size:
2 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15593371 2 sets 2 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 15593371 Supplier Invitrogen™ Supplier No. HC1001
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.

SureCast Glass Plates offer superior quality, durability and damage-resistance and are used to cast 1.0mm polyacrylamide gels using the SureCast Handcast Station. SureCast gels are designed for use in the Mini Gel Tank. SureCast Gel Spacers not included and are sold separately.

Product Features
  • Premium, ultra-durable glass plates
  • Damage-resistant design
  • Cast up to 20 times more gels using a set of glass plates versus main competitor's glass plates
  • Compatible only with the SureCast Handcast System

Recommended Storage

Room Temperature

Warranty

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 front plates and 2 back plates
For Use With (Equipment) SureCast Handcast System
Gel Size Mini
Product Line SureCast
Type SureCast™ Glass Plates
Quantity 2 sets
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.