missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Stacking Buffer

Product Code. 15558055
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
Pack of 2
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15558055 2 x 500 mL Pack of 2
15627968 5 x 500 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15558055 Supplier Invitrogen™ Supplier No. HC2112
  • ONLINE EXCLUSIVE PRICE
    36.08 €
    29.22 € / Pack of 2
    Save 6.86 €
    19% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 x 500 mL (1 L total) SureCast Stacking Buffer packs.

Store at room temperature.
Quantity 2 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Stacking Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.