missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureLock™ Tandem Midi Gel Tank Replacement Tank

Product Code. 17142514
Change view
Click to view available options
Quantity:
1 tank
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17142514 1 tank Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17142514 Supplier Invitrogen™ Supplier No. STM1003
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This tank is a replacement part for the SureLock Tandem Midi Gel Tank

About the SureLock Tandem Midi Gel Tank

The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SureLock Tandem Midi Gel Tank
Gel Size Midi
Product Line SureLock
Type Midi Gel Tank Replacement Tank
Quantity 1 tank
Shipping Condition Room Temperature
Warranty 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.