missing translation for 'onlineSavingsMsg'
Learn More

SureTect™ Salmonella species PCR Assay

Product Code. 17829142 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Food and Beverage
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17829142 Food and Beverage Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17829142 Supplier Thermo Scientific Supplier No. A56841
  • ONLINE EXCLUSIVE PRICE
    2810.00 €
    2005.16 € / Pack of 1
    Save 804.84 €
    28% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect Salmonella species in food and environmental samples with the Thermo Scientific™ SureTect™ Salmonella species PCR Assay.

Detect Salmonella species in food and environmental samples with the Thermo Scientific™ SureTect™ Salmonella species PCR Assay which incorporates optimized primer and probe components for highly-specific detection of Salmonella species.

The assay's workflows have been validated for a broad range of human foods, pet food, including large test portions, and environmental samples by AOAC International and ISO 16140-2:2016 according to NF VALIDATION™ by AFNOR Certification and by MicroVal™ Certification. The assay is also certified for primary production samples by NF Validation™ by AFNOR Certification.

The new SureTect assay format incorporates color-coded plastics with orientation markings and breakable strip format and colored reagents and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.

The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.

Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 10 to 24 hours
  • Maximum flexibility for operational efficiency
  • Run all assays simultaneously in the same run
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps
  • Detect multiple targets in the same run with universal PCR protocols across the range

Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results
  • NOW available with SureClean™ for your most challenging samples where free DNA can influence result performance

The Thermo Scientific SureTect Salmonella species PCR Assay Kit includes:

  • Lysis Reagent 1 Tubes
  • Proteinase K
  • Salmonella species PCR Assay Tubes
  • Thermo Scientific 96-Well Pierceable Seals
  • PCR Caps

The SureTect Salmonella PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System using RapidFinder™ Analysis Software v3.0 or later or the Applied Biosystems™ 7500 Fast Food Safety Real-Time PCR Instrument using Rapidfinder™ Express Software v2.0 or later, allowing you to test easily and confidently. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.

The SureTect Salmonella PCR Assay has been tested extensively on a wide range of human food categories, including meat products, dairy products, fish and fishery products, fruits, and vegetables. The assay has also been tested on pet food and environmental samples, including primary products samples. Individual matrices tested in the method validation studies were selected based on their associated contamination by Salmonella and the known challenges linked with testing them by PCR methods. Individual matrices included but were not limited to ground beef, raw poultry, eggs, non-fat dried milk, and ready-to-eat meals.

TRUSTED_SUSTAINABILITY

Specifications

Product Type SureTect Salmonella spp
For Use With (Application) Food and Beverage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.