missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ SURFA'SAFE 02

Product Code. 13486298
Change view
Click to view available options
Volume (Metric):
750 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13486298 750 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13486298 Supplier Laboratoires Anios™ Supplier No. 2178544

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratoires Anios™ SURFA'SAFE 02 is a natural and biodegradable broad spectrum antimicrobial spray.

  • Contains 100% easily biodegradable organic components
  • Vegetable sourced ethanol
  • Broad antimicrobial spectrum
  • Broad material compatibility
  • Suitable for cleaning and disinfecting medical devices and surfaces in operating theaters, high risk services, care services
  • Dispensed foam limits formation of aerosols and volatile components
  • Bactericidal, tuberculocide, fungicide, virucide

Specifications

Product Type Disinfectant
Certifications/Compliance Ecodétergent certified by ECOCERT Greenlife.Bactericidal: EN1040, EN13727, EN13697.Tuberculocide: EN14348, EN13697.Fungicide: EN1275, EN 13624, EN13697.Active according EN14476 + A1 on PRV (model virus HBV), BVDV (of HCV), Vaccinia virus, herpesvirus, rotavirus
Composition 30%: water, disinfectant (s): Ethanol (CAS No. 64-17-5: 380mg/g), hydrogen peroxide (CAS No. 7722-84
For Use With (Application) Cleaning and disinfecting medical devices and surfaces in operating theaters, high risk services, care services
Quantity 12/Cs.
Volume (Metric) 750 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.