missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Surface pH Electrodes

Product Code. 11796338 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
12 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11796338 12 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11796338

Please to purchase this item. Need a web account? Register with us today!

Surface pH electrode that is ideal for measurements on paper, skin, leather, agar, and electrophoresis gel.

  • pH electrode in flat glass 12 or 8 mm
  • For measurements on paper, skin, leather, agar, electrophoresis gel

Specifications

Description Surface pH Electrodes
Diameter (Metric) 12 mm
Type pH Electrodes, 12 mm
For Use With (Application) Surface pH electrode that is ideal for measurements on paper, skin, leather, agar, and electrophoresis gel.
pH Range 0 to 12
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.