missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Surfanios IP Sterile PAE

Product Code. 15461243
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15461243 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461243 Supplier Laboratoires Anios™ Supplier No. 2409534FJ
  • ONLINE EXCLUSIVE PRICE
    455.70 €
    403.87 € / Pack of 6
    Save 51.83 €
    11% Off
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratoires Anios™ Surfanios IP Sterile PAE is a broad spectrum biocidal suitable for cleaning and disinfection of floors, walls, surfaces and equipment in the pharmaceutical industry for clean rooms (A and B grades).

  • Ready to use biocidal product with broad spectrum
  • Synergistic combination of 2 antimicrobial active ingredients
  • Aldehyde free
  • Activity in compliance with European standards
  • Made with WFI
  • Double-bagged packaging
  • Gamma-irradiated

Specifications

Product Type Disinfectant Spray
pH 9 to 12 (at 20°C)
Certifications/Compliance Bactericide: N 1276, EN 13697, Yeasticide: EN 1650
Composition N-(3-aminopropyl)-N-dodecylpropan-1,3-diamine -N° CAS 2372-82-9 (0,51 mg/g), Didecyldimethylammo-nium chloride (N° CAS 7173-51-5 (0,25mg/g).
Container Type Spray Bottle
For Use With (Application) Surface and equipment disinfection in clean rooms (A and B)
Quantity 6/Cs.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.