missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Surfanios Premium
Description
Detergent disinfectant for floors and surfaces:
- Green coloured solution
- pH of pure product: 11.2
- pH of diluted product (0.25%): approx 8
- Density: 1.007
- Refraction index: 1.3587
Specifications
Specifications
| Product Type | Surfanios Premium |
| Composition | N- (3- aminopropyl) -N- dodecylpropane -1,3- diamine, didecyldimethylammonium chloride, excipients |
| For Use With (Application) | For Cleaning and Disinfection of Floors and Surfaces |
| Includes | with Pump 20mL |
| Quantity | 4/Pk. |
| Volume (Metric) | 5 L |
Safety and Handling
Product Identifier
- SURFANIOS PREMIUM
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contain: N-(3-AMINOPROPYL)-N-DODECYLPROPANE-1, 3-DIAMINE, DIDECYLDIMETHYLAMMONIUM CHLORIDE, PROPAN-2-OL
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?