missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios Surfanios™ Premium Disinfectant Detergent

Product Code. 11756023
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11756023 1 L Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11756023 Supplier Laboratoires Anios Supplier No. 1917092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleaning and disinfection of floors, walls, medical equipment and non-invasive medical devices.

Detergent disinfectant for floors and surfaces:

  • Green coloured solution
  • pH of pure product: 11.2
  • pH of diluted product (0.25%): approx 8
  • Density: 1.007
  • Refraction index: 1.3587

Specifications

Form Liquid
pH 10.9
Certifications/Compliance EN 1040, EN 1327 and 1276, EN 1275, EN 1650, EN 14348
Content And Storage 5°C to 35°C
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • SURFANIOS PREMIUM
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P234-Keep only in original container.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: N-(3-AMINOPROPYL)-N-DODECYLPROPANE-1, 3-DIAMINE, DIDECYLDIMETHYLAMMONIUM CHLORIDE, PROPAN-2-OL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.