Learn More
Rotulex™ Metal Joint Clamps
Spring-loaded metal clips. Rotulex™ Metal Joint Clamps are designed to secure corresponding Rotulex™ glassware systems with ball and socket style joints.
25.15 € - 955.00 €
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13191364
|
SVL
RC35 |
Approx. bore size: 20 mm |
52.25 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
12059032
|
SVL
RC41 |
Approx. bore size: 25 mm |
70.85 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
12099111
|
SVL
RC29 |
Approx. bore size: 15 mm |
31.50 €
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
12434751
|
SVL
RC13 |
Approx. bore size: 5 mm |
25.15 €
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
15557950
|
SVL
RC64 |
Approx. bore: 40 mm |
955.00 €
Pack of 5 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
10686475
|
SVL
RC19 |
Approx. bore: 9 mm |
28.55 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- Suitable for high temperature work
- System includes male ball joint, PTFE covered O-ring and female cup joint which are secured with the joint clamp
- Components are ordered separately
- Available in a variety of sizes