Learn More
SVL™ PTFE Key
Description
- For use with right angle pattern Torion stopcock
- Screwed into the threaded Torion connector, to open and close the stopcock
Specifications
Specifications
| Type | Stopcock Key |
| Material | PTFE |
| For Use With (Equipment) | Torion Stopcock 722-43 |
Product Title
Haben Sie Verbesserungsvorschläge?