Learn More
SVL™ Sealing Ring for Butt Joint
Product Code. 10580291
Change view
Click to view available options
Joint Size:
15
22
30
42
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size unitSize
10580291 22 Pack of 10
12691591 30 Pack of 10
12681591 15 Pack of 10
12093948 15 Each
12977016 42 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10580291 Supplier SVL Supplier No. 70102
Add to basket
Add to basket
Manufactured from silicone rubber
  • Silicone rubber and sheathed with PTFE for protection against chemical attack
  • Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
TRUSTED_SUSTAINABILITY
Specifications
Description Joint size: 22
For Use With (Application) Butt joints
Joint Size 22
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring