missing translation for 'onlineSavingsMsg'
Learn More

SVL™ Sealing Rings for Sliding Joints

Product Code. 12607245
Change view
Click to view available options
Diameter (Metric) Inside:
11.6 to 12.4 mm
13.6 to 14.4 mm
15.6 to 16.4 mm
17.6 to 18.4 mm
19.5 to 20.5 mm
21.5 to 22.5 mm
29.2 to 30.8 mm
31.2 to 32.8 mm
4.0 to 4.4 mm
5.6 to 6.4 mm
6.6 to 7.4 mm
7.6 to 8.4 mm
9.6 to 10.4 mm
Unit Size:
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
12607245 13.6 to 14.4 mm Pack of 10
10105674 6.6 to 7.4 mm Pack of 10
12517816 17.6 to 18.4 mm Pack of 10
10287774 31.2 to 32.8 mm Pack of 10
10739365 21.5 to 22.5 mm Pack of 10
10442205 7.6 to 8.4 mm Pack of 10
10635905 4.0 to 4.4 mm Pack of 10
10645905 5.6 to 6.4 mm Pack of 10
10146444 15.6 to 16.4 mm Pack of 10
10319944 9.6 to 10.4 mm Pack of 10
10379984 11.6 to 12.4 mm Pack of 10
10014464 19.5 to 20.5 mm Pack of 10
11943889 29.2 to 30.8 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 12607245 Supplier SVL Supplier No. 70124
  • ONLINE EXCLUSIVE PRICE
    137.50 €
    108.66 € / Pack of 10
    Save 28.84 €
    20% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion. SVL™ Sealing Rings for Sliding Joints are ideal for inserting thermometers and glass tubing into an SVL™ system. A wide range of sizes is available for use with tubing from 4.0 to 32.8mm.

  • Sealing rings designed to provide a sliding joint
  • Ideal for inserting thermometers and glass tubing etc. into an SVL™ system
  • Sealing rings manufactured from silicone rubber and sheathed with PTFE for protection against chemical erosion
  • Wide range of sizes for use with tubing from 4.0 to 32.8mm

Specifications

Diameter (Metric) Inside 13.6 to 14.4 mm
For Use With (Application) Torion joints and sliding joints
For Use With (Equipment) Glass Body 721-03
Joint Size 22
Material Silicone Rubber, PTFE (Sheath)
Type Seal Ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.