missing translation for 'onlineSavingsMsg'
Learn More

SVOC Mixture 506 2000 μg/mL in Acetone:Hexane, Dr. Ehrenstorfer

Product Code. 30325225
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325225 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325225 Supplier Dr. Ehrenstorfer Supplier No. DREA50000506AH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluorobiphenyl∣p-Terphenyl D14
CAS 321-60-8, 1718-51-0
Solvent or Matrix Acetone/Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2-Fluorobiphenyl 2000 μg/mL∣p-Terphenyl D14 2000 μg/mL
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.