Learn More
3M™ Swab-Sampler
Convenient and quick collection device for environmental sampling and testing over small surface areas.
Brand: 3M™ 7000053818
Additional Details : CAS Number : 7732-18-5 Weight : 0.34500kg
Description
- Ready to use for pathogen testing of environmental samples.
- Swab reaches into corners and hard-to-reach areas for increased accuracy.
- Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
- Bottles have ample headspace for sample addition and vortexing.
- Self-contained swab and vial for increased productivity and efficiency.
Specifications
10 mL Neutralizing Buffer | |
100 | |
Swab |