Learn More
Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Product Code. 15297488 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
3-Port SilFlow MCD Device
Swap-IT
Description:
Blanking Ferrule
Chromatography Accessories
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Description Unit Size
15297488 3-Port SilFlow MCD Device Blanking Ferrule Pack of 5
11567761 Swap-IT Chromatography Accessories Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15297488 Supplier Thermo Scientific™ Supplier No. 290ST414
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
TRUSTED_SUSTAINABILITY
Specifications
Description Blanking Ferrule
For Use With (Equipment) 3-Port SilFlow MCD Device