Learn More
Applied Biosystems™ SwiftArrayStudio™ Server
Description
The SwiftArrayStudio Server is a centralized management system designed to control and monitor individual SwiftArrayStudio microarray analyzer systems or fleets of up to four instruments simultaneously. This server platform allows comprehensive instrument status monitoring capabilities while offering integrated error detection and troubleshooting assistance to optimize laboratory workflow efficiency.
Key technical features include easy file upload functionality, remote monitoring capabilities, and intelligent diagnostic support through its Smart help system. The SwiftArrayStudio server's scalable architecture supports laboratory configurations ranging from single-instrument setups to multi-instrument, accommodating up to four SwiftArrayStudio microarray analyzer systems within a unified management framework.
Features
- Run Management: Comprehensive workflow control and execution monitoring capabilities for scientific experiments and analytical procedures
- Data Storage: Robust data repository system for secure storage, organization, and retrieval of experimental datasets and analytical results
- Instrument Management: Integrated control and monitoring of laboratory instrumentation with real-time status tracking and configuration management
- Data Analysis: Advanced analytical processing tools for statistical evaluation, visualization, and interpretation of experimental data
SwiftArrayStudio Server: Comprehensive Data Management and Analysis Platform
Automated Analysis Pipeline Accelerates Genotyping Workflows
The SwiftArrayStudio Server integrates Applied Biosystems' Automated Axiom™ Analysis suite to provide comprehensive quality control analysis and interpretation of genotyping microarray experiments. This automated integration streamlines the data analysis pipeline by automatically generating quality control reports for each array plate run, eliminating manual processing bottlenecks that traditionally slow research progress. The system leverages default Best Practices parameters to automatically generate genotype reports, significantly accelerating the rate at which researchers can obtain results and derive meaningful insights from their experimental data. This automation is particularly valuable given that microarray experiments generate substantial volumes of data that require systematic processing and analysis.
Centralized Data Management Enhances Laboratory Efficiency
The SwiftArrayStudio Server addresses the critical need for centralized data storage and management in high-throughput genotyping environments. By providing a single location for collecting and storing raw data from microarray experiments, the system enhances operational efficiency for both individual genotyping projects and entire laboratory operations. The server facilitates comprehensive management of large datasets generated from multiple microarray plates, ensuring that valuable experimental data remains organized, accessible, and secure throughout the research process.
Comprehensive Array Plate Registration Ensures Data Integrity
Efficient microarray experimentation requires meticulous registration of array plates to specify running conditions and establish clear data linkage from sample input to final scan output. The SwiftArrayStudio Server supports comprehensive registration processes that capture essential metadata including sample names, array barcodes, reagent lot information, array processing variables, and other critical parameters necessary for proper genotyping project management. This systematic approach to data registration ensures experimental traceability and maintains the integrity of the analytical chain from sample preparation through final results generation.
Real-Time Instrument Monitoring Optimizes Laboratory Operations
High-throughput genotyping laboratories require continuous monitoring of instrument performance to ensure consistent operation and high-quality results. The SwiftArrayStudio Server delivers advanced monitoring capabilities that provide real-time updates on plate and run status. This proactive approach enables laboratory personnel to quickly determine sample progress within a run and closely track results as they are generated, supporting reliable and reproducible outcomes.
Integrated Solution Delivers Unprecedented Throughput
The SwiftArrayStudio Server functions as an integral component of Applied Biosystems' comprehensive genotyping solution, designed for seamless integration with SwiftArrayStudio instrumentation. This integrated approach represents Applied Biosystems' continued innovation in high-throughput microarray technologies, enabling the SwiftArrayAssay and SwiftArrayStudio system to deliver complete genotyping information from a single array plate in just 30 hours. The server's role in facilitating efficient instrument and data management further accelerates the pace at which laboratories can progress from sample processing to meaningful scientific discoveries, establishing new benchmarks for productivity in genomic research environments.
Warranty
1 Year
Specifications
Specifications
| Product Type | Data Collection server (DCS) |
| For Use With (Equipment) | SwiftArrayStudio Microarray Analyzer |
| For Use With (Application) | Microarray |