missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Swing-bucket Rotor for SpinPro™ 8 R / Centrifuge CR22N

Product Code. 32226679 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
4 x 750 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
32226679 4 x 750 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32226679 Supplier Eppendorf Supplier No. 5720411482
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Offers a wide variety of rotors for High-Speed Centrifuges, ensuring the perfect fit for specific applications.

  • Rotor type: swing-bucket rotor
  • Application: For harvesting of bacteria, cells or yeast for e.g. recombinant protein or antibody purification in bottles with small to medium volumes
  • Max. speed: 4830 x g (4700 rpm)
  • Max. capacity: 4 x 750 mL (Nominal capacity)
  • Made from high-quality: aluminum
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Swinging Bucket Rotor
Capacity 4 x 750 mL
Max. RCF 4830 x g
Max. Speed 4700 rpm
K Factor 2354 (1.5 mL reaction vessel) to 10,562 (500 mL Corning bottle)
Biocontainment No
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.