missing translation for 'onlineSavingsMsg'
Learn More

mb2 Swivel Anti-Static Work Chair in Polyurethane - Permanent Contact - Glides

Product Code. 30130619
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30130619 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130619 Supplier mb2 Supplier No. 245B2C1D2E3
  • ONLINE EXCLUSIVE PRICE
    481.00 €
    421.34 € / Pack of 1
    Save 59.66 €
    12% Off
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Swivel antistatic work chair in polyurethane, permanent contact, middle high gas lift with footrest and glides

  • Antistatic work chair
  • Seat and backrest in multilayers of beech and poplar wood, entirely foamed with black antistatic polyurethane
  • Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
  • Gas height adjustment
  • Antistatic glides
  • 5-spoke base in die-cast polished aluminium

Specifica

Product Type ESD Chair
Color Black
Adjustable (Height) Yes
Autoclavable Not Autoclavable
Back Style Ergonomic
Certifications/Compliance IEC 61340-5-1
For Use With (Application) Laboratory
Lift Type Pneumatic
Quantity 1 Ea.
Seating Height 460 to 650 mm
Warranty 5 years for normal use
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support No
With Tilt Yes
Material Polyurethane
Seating Type Medium Bench Height
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.