missing translation for 'onlineSavingsMsg'
Learn More

mb2 Swivel Anti-Static Work Chair in Polyurethane

Product Code. 30130610
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30130610 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130610 Supplier mb2 Supplier No. 231B2C1D2E3
  • ONLINE EXCLUSIVE PRICE
    429.00 €
    414.52 € / Pack of 1
    Save 14.48 €
    3% Off
View all promotions
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Swivel antistatic work chair in polyurethane, middle high gas lift with footrest and glides

  • Antistatic work chair
  • Seat and backrest in multilayers of beech and poplar wood, entirely foamed with black antistatic polyurethane
  • Backrest support in chromed steel rod, in height and depth adjustable
  • Gas height adjustment
  • Antistatic glides
  • 5-spoke base in die-cast polished aluminium

Specifications

Product Type ESD Chair
Color Black
Adjustable (Height) Yes
Autoclavable Not Autoclavable
Back Style Standard
Certifications/Compliance IEC 61340-5-1
For Use With (Application) Laboratory
Lift Type Pneumatic
Quantity 1 Ea.
Seating Height 460 to 650 mm
Warranty 5 years for normal use
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Medium Bench Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.